missing translation for 'onlineSavingsMsg'
Learn More

Mouse Meteorin-like/METRNL PE-conjugated Antibody, R&D Systems™

Catalog No. IC6679P025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6679P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC6679P025 Supplier Bio-Techne Supplier No. IC6679P025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

Meteorin-like/METRNL Monoclonal specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen Meteorin-like/METRNL
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 829535
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8VE43
Gene Alias Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788
Gene Symbols METRNL
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-S-derived recombinant mouse Meteorin-like/METRNL Gln46-Glu311 Accession # Q8VE43
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284207
Test Specificity Detects mouse Meteorin-like/METRNL in direct ELISA.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Supernatant
Isotype IgG2b
Product Line R&D Systems
Show More Show Less