missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse LIGHT, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF315121MG
Encompass
Change view
Click to view available options
Quantity:
100 μg
20 μg
500 μg
1 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF315121MG 1 mg
AF31512100 100 μg
AF3151220UG 20 μg
AF31512500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AF315121MG Supplier Gibco™ Supplier No. AF315121MG

Recombinant Protein

Recombinant Murine LIGHT is a 20.1 kDa protein containing 183 amino acid residues, comprising the TNF-homologous region of the LIGHT extracellular domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.

Specifications

Accession Number Q9QYH9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 50930
Molecular Weight (g/mol) 20.1 kDa
Name Mouse LIGHT, Animal-Free
Quantity 1 mg
Source E. coli
Endotoxin Concentration <0.1 EU/ μg
Gene Alias CD258; Herpes virus entry mediator ligand; herpesvirus entry mediator ligand; HVEML; HVEM-L; Light; LTg; Ly113; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tnfsf14; TR2; tumor necrosis factor (ligand) superfamily, member 14; tumor necrosis factor ligand 1 D; tumor necrosis factor ligand superfamily member 14; Tumor necrosis factor ligand superfamily member 14, membrane form; Tumor necrosis factor ligand superfamily member 14, soluble form; tumor necrosis factor superfamily member 14; UNQ391/PRO726
Common Name LIGHT
Gene Symbol TNFSF14
Conjugate Unconjugated
Recombinant Recombinant
Sequence MRLHQRLGDI VAHLPDGGKG SWEKLIQDQR SHQANPAAHL TGANASLIGI GGPLLWETRL GLAFLRGLTY HDGALVTMEP GYYYVYSKVQ LSGVGCPQGL ANGLPITHGL YKRTSRYPKE LELLVSRRSP CGRANSSRVW WDSSFLGGVV HLEAGEEVVV RVPGNRLVRP RDGTRSYFGA FMV
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less