missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse LIF, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF250021MG
Encompass
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
1 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF250021MG 1 mg
AF25002100 100 μg
AF2500225UG 25 μg
AF25002500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AF250021MG Supplier Gibco™ Supplier No. AF250021MG

Recombinant Protein

Recombinant Murine LIF is a 19.9 kDa protein containing 180 amino acids residues, including three disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Leukemia inhibitory factor (LIF) is a 20 kDa protein that belongs to the IL-6 receptor family. It binds to a heterodimeric membrane receptor made up of a LIF-specific subunit, gp190 or LIFR, and the subunit gp130, which is shared with the other members of the IL-6 family. LIF expression has been observed in various tissues including thymus, lung, and neuronal tissue. LIF can be up-regulated by pro-inflammatory cytokines such as TNFα and IL-17, and elevated levels of LIF have been found in cases of rheumatoid arthritis, neural injury, systemic inflammation, and tuberculosis. LIF displays diverse biological effects, but is best known for its ability to inhibit the differentiation of embryonic stem cells in mice and contribute to stem cell self-renewal.Human and mouse LIF share 79% sequence homology and exhibit cross-species activity. However, LIF inhibition of stem cell differentiation appears to be mouse-specific.

Specifications

Accession Number P09056
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 16878
Molecular Weight (g/mol) 19.6 kDa
Name Mouse LIF, Animal-Free
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias CDF; cholinergic differentiation factor; cholinergic neuronal differentiation factor; d factor; DIA; differentiation inhibitory activity; differentiation-inducing factor; differentiation-stimulating factor; Emfilermin; hepatocyte-stimulating factor III; HILDA; human interleukin in DA cells; leukemia inhibitory factor; leukemia inhibitory factor (cholinergic differentiation factor); LIF; Melanoma-derived LPL inhibitor; MLPLI; myeloid leukaemia inhibitory factor
Common Name LIF
Gene Symbol Lif
Biological Activity PeproTech's Murine LIF is fully biologically active when compared to standards. The ED50 as determined by the M1 cell differentiation assay is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 10^7 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence SPLPITPVNA TCAIRHPCHG NLMNQIKNQL AQLNGSANAL FISYYTAQGE PFPNNVEKLC APNMTDFPSF HGNGTEKTKL VELYRMVAYL SASLTNITRD QKVLNPTAVS LQVKLNATID VMRGLLSNVL CRLCNKYRVG HVDVPPVPDH SDKEAFQRKK LGCQLLGTYK QVISVVVQAF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less