missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-9 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Catalog No. IC409N100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC409N100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC409N100UG Supplier Bio-Techne Supplier No. IC409N100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

IL-9 Monoclonal specifically detects IL-9 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-9
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 222604
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P15247.1
Gene Alias Cytokine P40, HP40, IL9, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40
Gene Symbols IL9
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-9 Gln19-Pro144 Accession # P15247.1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3578
Test Specificity Detects mouse IL-9 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human IL-9 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2a
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)