Learn More
Invitrogen™ Mouse IL-5 Uncoated ELISA Kit with Plates
ELISA
Supplier: Invitrogen™ 88705476
Description
The Mouse Interleukin-5 (IL-5) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-5. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.Specifications
| P04401 | |
| 4 pg/mL | |
| ELISA | |
| Mouse | |
| Colorimetric Microplate Reader | |
| GDCF-2,HC11,HSMCR30,MCAF,MCP-1,MCP1,SCYA2,SMC-CF | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| IL-5 | |
| 24 hrs 30 mins |
| 4 to 500 pg/mL | |
| HRP | |
| Lavage Fluid, Serum, Supernatant | |
| ELISA | |
| 16191 | |
| Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86) | |
| 20 x 96 Tests | |
| Lavage Fluid 100 μL, Serum 100 μL, Supernatant 100 μL | |
| Interleukin-5 | |
| 1 hrs 45 mins |