missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-27 PE-conjugated Antibody, R&D Systems™

Catalog No. IC1834P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1834P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC1834P Supplier Bio-Techne Supplier No. IC1834P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

IL-27 Monoclonal specifically detects IL-27 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-27
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 234205
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O35228 (EBI3) & Q8K3I6 (p28)
Gene Alias IL27, IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A
Gene Symbols IL27
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse IL-27 Tyr19-Pro228 (EBI3) & Phe29-Ser234 (p28) Accession # O35228 (EBI3) & Q8K3I6 (p28)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246778
Test Specificity Detects mouse IL-27 in Western blots. This antibody recognizes the p28 subunit either as part of a p28/EBI-3 heterodimer or as a free subunit.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line R&D Systems
Show More Show Less