missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-27 Alexa Fluor 594-conjugated Antibody, R&D Systems™
Description
IL-27 Monoclonal specifically detects IL-27 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | IL-27 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 234205 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 594 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | O35228 (EBI3) & Q8K3I6 (p28) |
| Gene Alias | IL-27, IL27 p28, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, IL-30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A |
| Show More |
For Research Use Only (RUO)