missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-12 p70 ELISA Kit

ELISA

$902.00 - $6,630.00

Specifications

Accession Number P43431,P43432
Assay Range 15.6 to 1,000 pg/mL
Assay Sensitivity 10.0 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-12 p70
Biotin-Conjugate anti-mouse IL-12 p70 monoclonal antibody
Streptavidin-HRP
Mouse IL-12 p70 Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
5018270
View Documents Promotion Details
Invitrogen™
BMS6004
96 Tests
Each for $902.00
Only null left
 
5018271
View Documents Promotion Details
Invitrogen™
BMS6004TEN
10 x 96 Tests
Each for $6,630.00
Only null left
 
Description

Description

The Mouse Interleukin-12 p70 (Ms IL-12 p70) ELISA quantitates Ms IL-12 p70 in mouse serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-12 p70. Principle of the method The Mouse IL-12 p70 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
Specifications

Specifications

P43431,P43432
10.0 pg/mL
ELISA
Mouse
Colorimetric Microplate Reader
IL-12p35,Il-12 A,Ll12a,p35,Il-12 b,Il-12p40,Il12p40,p40
8.3
HRP
Serum 50 μL, Supernatant 50 μL
Interleukin-12 subunit p70, CLMF, NKSF
1 hrs 20 mins
15.6 to 1,000 pg/mL
HRP
Serum, Supernatant
ELISA
16159, 16160
11%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
IL-12 p70
3 hrs 10 mins
Videos
SDS
Documents

Documents