missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse GM-CSF ELISA Kit

ELISA

$908.00

Specifications

Accession Number P01587
Assay Range 3.9-250 pg/mL
Assay Sensitivity 2.0 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse GM-CSF
Biotin-Conjugate anti-mouse GM-CSF monoclonal antibody
Streptavidin-HRP
Mouse GM-CSF Standard lyophilized, 500pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
5018320
View Documents Promotion Details
Invitrogen™
BMS612
96 Tests
Each for $908.00
Only null left
 
Description

Description

The Mouse Granulocyte Macrophage Colony Stimulating Factor (Ms GM-CSF) ELISA quantitates Ms GM-CSF in mouse serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms GM-CSF. Principle of the method The Mouse GM-CSF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
Specifications

Specifications

P01587
2.0 pg/mL
ELISA
Mouse
Absorbance Microplate Reader
CSF2
5%
Biotin
Serum 50 μL, Supernatant 50 μL
Molgramostin, Sargramostim
1 hr 20 min
3.9-250 pg/mL
HRP
Serum, Supernatant
ELISA
12981
10%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2-8°C
GM-CSF
3 hr 30 min
Videos
SDS
Documents

Documents