missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; Ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; Tumor antigen HOM-HD-21; UAT; Urate transporter/channel; urate transporter/channel protein |
|---|---|
| Assay Range | 0.18 to 40 ng/mL |
| Assay Sensitivity | 0.18 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EM33RBX5
|
Invitrogen™
EM33RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EM33RB
|
Invitrogen™
EM33RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Mouse Galectin-9 quantitates mouse Galectin-9 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Galectin-9.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
| 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; Ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; Tumor antigen HOM-HD-21; UAT; Urate transporter/channel; urate transporter/channel protein | |
| 0.18 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Lgals9 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Galectin 9 | |
| 4 hrs 45 mins |
| 0.18 to 40 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 16859 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |