missing translation for 'onlineSavingsMsg'
Learn More

Mouse Galectin-9 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Catalog No. IC3535U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3535U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC3535U100 Supplier Bio-Techne Supplier No. IC3535U100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

Galectin-9 Monoclonal specifically detects Galectin-9 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen Galectin-9
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 766428
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O08573-2
Gene Alias ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Gene Symbols LGALS9
Host Species Rat
Immunogen E. coli-derived recombinant mouse Galectin-9 (short isoform) Ala2-Thr322 Accession # O08573-2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3965
Test Specificity Detects mouse Galectin-9 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Galectin-9 or recombinant mouse Galectin-4 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)