missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Dtk ELISA Kit

Catalog No. EM22RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM22RBX5 5 x 96 Tests
EM22RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EM22RBX5 Supplier Invitrogen™ Supplier No. EM22RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

Mouse Dtk quantitates mouse Dtk in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Dtk.

TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, signal transduction, cell adhesion. and cytoskeleton reorganization.

Specifications

Accession Number P55144
Assay Range 0.4-100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; Dtk; Etk-2; etk2/tyro3; FLJ16467; Rek; Rse; Sky; soluble tyro 3; soluble tyro3; styro 3; styro3; TIF; TK19-2; tyro 3; TYRO3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase receptor TYRO3; Tyrosine-protein kinase RSE; tyrosine-protein kinase SKY; Tyrosine-protein kinase TIFAI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; Dtk; Etk-2; etk2/tyro3; FLJ16467; Rek; Rse; Sky; soluble tyro 3; soluble tyro3; styro 3; styro3; TIF; TK19-2; tyro 3; TYRO3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase receptor TYRO3; Tyrosine-protein kinase RSE; tyrosine-protein kinase SKY; Tyrosine-protein kinase TIF;
Gene ID (Entrez) 22174
Gene Symbol TYRO3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target TYRO3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less