Learn More
Invitrogen™ Mouse CTX ELISA Kit
Description
Mouse CTX ELISA Kit quantifies CTX in serum and plasma. The Mouse CTX solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Specifications
Specifications
| Assay Range | 31.25-2000 pg/mL |
| Assay Sensitivity | 18.75 pg/mL |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| Target Species | Mouse |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | C-terminal telopeptide antibody; C-Terminal Telopeptide of Type I CollagenC-terminal telopeptide antibody; C-Terminal Telopeptide of Type I Collagen; |
| Show More |