missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Creatine Kinase MM ELISA Kit

Catalog No. EEL217
Change view
Click to view available options
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EEL217 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL217 Supplier Invitrogen™ Supplier No. EEL217

ELISA

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.

Specifications

Accession Number P07310
Assay Range 1.56 to 100 ng/mL
Assay Sensitivity 0.94 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format ELISA Microplate (Dismountable)
Product Type ELISA
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 12715
Gene Symbol CKM
Interassay CV <10
Intraassay CV <10
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Creatine Kinase MM
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less