missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Collagen I ELISA Kit

Catalog No. EEL218
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL218 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL218 Supplier Invitrogen™ Supplier No. EEL218

ELISA

Mouse Collagen I ELISA Kit quantifies Collagen I in serum and plasma. The Mouse Collagen I solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Collagen I is a Type I collagen with a triple helix structure comprised of two alpha-1 chains and one alpha-2 chain. Collagen I is a member of group I collagen (fibril-forming collagen) found in most connective tissues, and is abundant in bone, cornea, dermis and tendon. Mutations in the COL1A2 gene encoding the alpha-2 chain are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in the COL1A2 gene which codes for the alpha-2 chain, however, tend to be less severe than mutations in the COL1A1 gene, reflecting the different role of alpha-2 chains in matrix integrity. Reciprocal translocations between chromosomes 17 and 22, where Collagen Type 1 genes and the gene for platelet derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified.

Specifications

Accession Number P11087, Q01149
Assay Range 0.31-20 ng/mL
Assay Sensitivity 0.19 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AA960264; AI325291; alpha 1 type I collagen; alpha 2 type I collagen; alpha 2 type I procollagen; alpha 2(I) collagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 chain of type I collagen; alpha-1 collagen (I); alpha-1 type 1 collagen; alpha-1 type I collagen; alpha-1 type-1 collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; alpha-2 type I collagen; alpha-2(1) collagen; alpha2(I) collagen; COL1; Col1a1; Col1a-1; COL1A1; matrix protein; Col1a2; Col1a-2; COLA; Cola1; Cola-1; Cola2; Cola-2; COLIA1; collagen; collagen 1 alpha 2 chain; collagen a2; collagen alpha 1 chain type I; collagen alpha 2(I) chain precursor; collagen alpha-1 chain; collagen alpha-1 chain precursor; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen alpha-2; collagen alpha-2 chain precursor; collagen alpha-2(I) chain; collagen COL1A2; collagen I alpha-2 polypeptide; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen type I alpha 1 chain; collagen type I alpha 2 chain; collagen, type 1, alpha 1; collagen, type I, alpha 1; collagen, type I, alpha 2; EDSC; Mov13; Mov-13; OI1; OI2; OI3; OI4; oim; osteogenesis imperfecta; precollagen alpha-2; prepro-alpha-1 collagen type I; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; similar to pro alpha 1(I) collagen; Type I Collagen; type I collagen alpha 1 chain; type I collagen alpha1; type I collagen alpha-2 chain; type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I-alpha 1 collagenAA960264; AI325291; alpha 1 type I collagen; alpha 2 type I collagen; alpha 2 type I procollagen; alpha 2(I) collagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 chain of type I collagen; alpha-1 collagen (I); alpha-1 type 1 collagen; alpha-1 type I collagen; alpha-1 type-1 collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; alpha-2 type I collagen; alpha-2(1) collagen; alpha2(I) collagen; COL1; Col1a1; Col1a-1; COL1A1; matrix protein; Col1a2; Col1a-2; COLA; Cola1; Cola-1; Cola2; Cola-2; COLIA1; collagen; collagen 1 alpha 2 chain; collagen a2; collagen alpha 1 chain type I; collagen alpha 2(I) chain precursor; collagen alpha-1 chain; collagen alpha-1 chain precursor; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen alpha-2; collagen alpha-2 chain precursor; collagen alpha-2(I) chain; collagen COL1A2; collagen I alpha-2 polypeptide; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen type I alpha 1 chain; collagen type I alpha 2 chain; collagen, type 1, alpha 1; collagen, type I, alpha 1; collagen, type I, alpha 2; EDSC; Mov13; Mov-13; OI1; OI2; OI3; OI4; oim; osteogenesis imperfecta; precollagen alpha-2; prepro-alpha-1 collagen type I; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; similar to pro alpha 1(I) collagen; Type I Collagen; type I collagen alpha 1 chain; type I collagen alpha1; type I collagen alpha-2 chain; type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I-alpha 1 collagen;
Gene ID (Entrez) 12842, 12843
Gene Symbol COL1A1, COL1A2
Interassay CV <10%
Intraassay CV <10%
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard , Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis, Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Shipping Condition Wet Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Collagen I
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less