missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | Chd; Chordin; Chrd; UNQ217/PRO243 |
|---|---|
| Assay Range | 49 to 12,000 pg/mL |
| Assay Sensitivity | 49 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EM17RBX5
|
Invitrogen™
EM17RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EM17RB
|
Invitrogen™
EM17RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Mouse Chordin quantitates mouse Chordin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Chordin.
This gene encodes a secreted protein that dorsalizes early vertebrate embryonic tissues by binding to ventralizing TGF-beta-like bone morphogenetic proteins and sequestering them in latent complexes. The encoded protein may also have roles in organogenesis and during adulthood. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequence has not been determined.Specifications
| Chd; Chordin; Chrd; UNQ217/PRO243 | |
| 49 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| CHRD | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Chordin | |
| 4 hrs 45 mins |
| 49 to 12,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 12667 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |