missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIBCO™ Mouse (CD-1) Microsomes
Description
Liver microsomes prepared from a pool of male CD-1 mice.
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Link
Learn more about our microsomes and subcellular fractions
Specifications
Specifications
| Quantity | 0.5 mL |
| Purity | Research Grade |
| Host Species | Mouse |
| Concentration | 20 mg/mL |
| Product Type | Mouse Microsome |
| Content And Storage | Store between -60°C and -85°C. |
| Format | Tube |
| Sub Cellular Localization | Microsomes |
| Shipping Condition | Dry Ice |
| Product Line | Gibco |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.