missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CCL2 (MCP-1) Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $4890.00
Specifications
| Accession Number | P10148 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 20296 |
| Molecular Weight (g/mol) | 13.8 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
25010100UG
|
Gibco™
LS25010100UG |
100 μg |
Each for $978.00
|
|
|||||
2501010UG
|
Gibco™
LS2501010UG |
10 μg |
Each for $317.00
|
|
|||||
250101MG
|
Gibco™
LS250101MG |
1 mg |
Each for $4,890.00
|
|
|||||
25010250UG
|
Gibco™
25010250UG |
250 μg |
Each for $1,626.02
|
|
|||||
250102UG
|
Gibco™
LS250102UG |
2 μg |
Each for $132.00
|
|
|||||
25010500UG
|
Gibco™
25010500UG |
500 μg |
Each for $2,795.00
|
|
|||||
2501050UG
|
Gibco™
2501050UG |
50 μg |
Each for $555.97
|
|
|||||
Description
Recombinant Murine JE is a 13.8 kDa protein containing 125 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.Specifications
| P10148 | |
| protein with no preservative | |
| 13.8 kDa | |
| E. coli | |
| <1 EU/ μg | |
| CCL2 (MCP-1) | |
| Determined by its ability to chemoattract Balb/C mouse spleen MNCs using a concentration range of 1.0-20.0 ng/ml | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 20296 | |
| Mouse CCL2 (MCP-1) | |
| RUO | |
| Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1 A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1 A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF | |
| Ccl2 | |
| Unconjugated | |
| QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |