missing translation for 'onlineSavingsMsg'
Learn More

Mouse B7-H1/PD-L1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Catalog No. FAB9078R025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9078R025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB9078R025 Supplier Bio-Techne Supplier No. FAB9078R025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

PD-L1/B7-H1 Monoclonal specifically detects PD-L1/B7-H1 in Mouse samples. It is validated for Flow Cytometry∣Immunocytochemistry.

Specifications

Antigen PD-L1/B7-H1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 929903
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9EP73
Gene Alias Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Gene Symbols Cd274
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse B7-H1/PD-L1 Met1-Thr239 Accession # Q9EP73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29126
Test Specificity Detects mouse B7-H1/PD-L1 in direct ELISAs. Stains mouse B7-H1/PD-L1 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line R&D Systems
Show More Show Less