missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain, Mouse, Clone: CH15, Abnova™
Mouse monoclonal antibody raised against partial recombinant Kappa Light Chain.
Supplier: Abnova Corporation MAB9548
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:200) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 3 x 1 mL | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Immunohistochemistry | |
| CH15 | |
| Mouse monoclonal antibody raised against partial recombinant Kappa Light Chain. | |
| In tissue culture supernatant (0.09% sodium azide) | |
| Recombinant protein corresponding to 106 amino acids of human Kappa Light Chain. | |
| RUO | |
| Human | |
| Liquid |