missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Mounted Plane Mirrors

Silvered back with protective coating.

$15.25 - $16.18

Specifications

Product Type Mounted Plane Mirror
Lens Material Glass
For Use With (Application) Studying optics (the physics of light) in a visually and kinesthetically effective manner
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
S24354
View Documents Promotion Details
Eisco™
PH0515A
75 x 25 mm
Each for $15.25
Only null left
 
S24355
View Documents Promotion Details
Eisco™
PH0515B
75 x 50 mm
Each for $16.18
Only null left
 
Description

Description

  • Made from glass
  • Mounted on 20mm thick wooden block
Specifications

Specifications

Mounted Plane Mirror
Glass
Studying optics (the physics of light) in a visually and kinesthetically effective manner
Videos
SDS
Documents

Documents