missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Mortar & Pestle Set, Porcelain, Fully Glazed
United Scientific™ Mortar & Pestle Set feature deep form mortars with lips are fully glazed on the inside and outside.
$17.75 - $75.04
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S89635
|
United Scientific™
PMG080 |
80 mL |
Each for $17.75
|
|
|||||
|
S89637
|
United Scientific™
PMG300 |
300 mL |
Each for $47.29
|
|
|||||
|
S89638
|
United Scientific™
PMG500 |
500 mL |
Each for $75.04
|
|
|||||
Description
Deep form mortars with lips are fully glazed on the inside and outside. Oversized pestles are also fully glazed to grinding surface.
- Tolerance is ±5 mm
- Offered in 80 mL, 125 mL, 300 mL, and 500 mL Capacity
Specifications
| Mortar and Pestle | |
| Porcelain | |
| Glazed |