Learn More
Fisherbrand™ Mortar and Pestle Deep Form
Mortar and Pestle Deep Form
Supplier: Fisherbrand™ CD100
Description
- Low permeability
- High strength
- Hard
- High resistance to chemical attacks
- Translucent or glazed body
Specifications
| 100mm | |
| Agate Stone | |
| Mortar and Pestle |
| Mortar and Pestle | |
| For grinding or crushing |