missing translation for 'onlineSavingsMsg'
Learn More

Morpholinosulfur Trifluoride 93.0+%, TCI America™

Catalog No. M15731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15731G 1 g
M15735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15731G Supplier TCI America Supplier No. M15731G

Chemical Identifiers

CAS 51010-74-3
Molecular Formula C4H8F3NOS
Molecular Weight (g/mol) 175.169
MDL Number MFCD00037057
InChI Key UFXIRMVZNARBDL-UHFFFAOYSA-N
Synonym morpholinosulfur trifluoride, morpholinosulfurtrifluoride, morph-dast, 4-trifluoro-$l^ 4-sulfanyl morpholine, 4-trifluoro-??-sulfanyl morpholine, morpholinosulphur trifluoride, morpho-dast, acmc-209krh, trifluoro morpholin-4-yl, morpholinotrifluorosulfur iv
PubChem CID 2733258
IUPAC Name trifluoro(morpholin-4-yl)-$l^{4}-sulfane
SMILES C1COCCN1S(F)(F)F

Specifications

Color Yellow
Boiling Point 42°C
Quantity 1 g
UN Number 3265
Formula Weight 175.17
Percent Purity ≥93.0% (T)
Physical Form Liquid
Chemical Name or Material Morpholinosulfur Trifluoride
TSCA No
Recommended Storage Refrigerator