accessibility menu, dialog, popup

UserName

ONSFFF

CAS RN 51010-74-3

IUPAC Name: 4-(trifluoro-λ⁴-sulfanyl)morpholine
Synonyms: 4-(trifluoro-λ⁴-sulfanyl)morpholine
Molecular Weight (g/mol): 175.17
Molecular Formula: C4H8F3NOS
InChi Key: UFXIRMVZNARBDL-UHFFFAOYSA-N
SMILES: FS(F)(F)N1CCOCC1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)