Learn More
Morphix Technologies SafeAir™ Detection Badges
Single-level screening badges provide economical daily exposure monitoring.
Supplier:  Morphix Technologies 38200150
Encompass
 View more versions of this product
Catalog No. 19087519
Description
Description
When the target gas is detected at the threshold level, an immediate color change occurs and an exclamation point is visible within the color triangle on the badge.
  • Provide immediate results
  • Easy to read
  • No laboratory analysis is required
  • May be used for as little as 15 minutes or up to two days; for minimum/maximum recommended sampling times, exposure level, and minimum detectable limit for each analyte, contact your Fisher Safety Customer Service Representative
Order Info
Lightweight badges attach to user's collar with a clip (19-087-526, sold separately). Available 50 badges per pack.
Specifications
Chemical Identifiers
Personnel or area monitoring for exposure times ranging from 15 min. to 24 Hours
0.003 lb.
Detection Badge
0.4 ppm.Hour
0.001 kg
Specifications
Personnel or area monitoring for exposure times ranging from 15 min. to 24 Hours
0.4 ppm.Hour
15.55°C to 32.77°C
0.001 kg
Aromatic Isocyanates
60°F to 91°F
0.003 lb.
Detection Badge
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1 Year