missing translation for 'onlineSavingsMsg'
Learn More

Morphine Sulfate Salt Solution, 1.0 mg/mL in Methanol, Drug Standard, MilliporeSigma™ Supelco™

Catalog No. 111016605
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016605 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016605 Supplier Merck Emd Millipore Supplier No. M95241ML

Specifications

Packaging Ampule
CAS 64-31-3
Linear Formula C34H38N2O6 · H2SO4
Concentration 1.0 mg/mL in methanol
MDL Number MFCD00069309
Quantity 1 mL
Formula Weight 668.75
Grade Drug Standard