missing translation for 'onlineSavingsMsg'
Learn More

Mordant Blue 29, TCI America™

Catalog No. D024125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D024125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D024125G Supplier TCI America Supplier No. D024125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
Synonym mordant blue 29, chromeazurol s, chrome azurol s, trisodium 5-3-carboxylato-5-methyl-4-oxo-cyclohexa-2,5-dien-1-ylidene-2,6-dichloro-3-sulfonato-phenyl methyl-2-hydroxy-3-methyl-benzoate, trisodium 5-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene-2,6-dichloro-3-sulfonatophenyl methyl-2-hydroxy-3-methylbenzoate
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 605.28
Physical Form Crystalline Powder
Chemical Name or Material Mordant Blue 29
EINECSNumber (5)-2088
TSCA Yes