missing translation for 'onlineSavingsMsg'
Learn More

Mordant Blue 13, TCI America™

Catalog No. A058425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A058425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A058425G Supplier TCI America Supplier No. A058425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1058-92-0
Molecular Formula C16H9ClN2Na2O9S2
Molecular Weight (g/mol) 518.80
MDL Number MFCD00003941
InChI Key RAIRBURLMTXYPH-WTBAQZMLSA-L
Synonym mordant blue 13, disodium 3-2-5-chloro-2-hydroxyphenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonate
PubChem CID 44134900
IUPAC Name disodium (3Z)-3-[2-(5-chloro-2-hydroxyphenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].OC1=CC=C(Cl)C=C1N\N=C1\C(=O)C2=C(O)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Black-Yellow
Quantity 25 g
Formula Weight 518.80
Physical Form Crystalline Powder
Chemical Name or Material Mordant Blue 13
EINECSNumber (5)-2086
TSCA Yes