Learn More
MOPSO sodium salt, Thermo Scientific™
Chemical Identifiers
| CAS | 79803-73-9 |
|---|---|
| Molecular Formula | C7H14NNaO5S |
| Molecular Weight (g/mol) | 247.24 |
| InChI Key | WSFQLUVWDKCYSW-UHFFFAOYNA-M |
| IUPAC Name | sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate |
| SMILES | [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC462140250
|
Thermo Scientific Chemicals
462140250 |
25 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC462141000
|
Thermo Scientific Chemicals
462141000 |
100 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC462145000
|
Thermo Scientific Chemicals
462145000 |
500 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 79803-73-9 | |
| 247.24 | |
| sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate |
| C7H14NNaO5S | |
| WSFQLUVWDKCYSW-UHFFFAOYNA-M | |
| [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O |
Specifications
| 79803-73-9 | |
| 25 g | |
| WSFQLUVWDKCYSW-UHFFFAOYNA-M | |
| sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate | |
| 247.24 |
| C7H14NNaO5S | |
| Solubility aq. soln.: 500 g/L | |
| [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O | |
| 247.24 | |
| MOPSO sodium salt |
Safety and Handling
GHS H Statement Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only