Learn More
MOPSO sodium salt, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 462141000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| MOPSO sodium salt | |
| C7H14NNaO5S | |
| Solubility aq. soln.: 500 g/L | |
| [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O | |
| 247.24 |
| 79803-73-9 | |
| 100 g | |
| WSFQLUVWDKCYSW-UHFFFAOYNA-M | |
| sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate | |
| 247.24 |
Chemical Identifiers
| 79803-73-9 | |
| 247.24 | |
| sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate |
| C7H14NNaO5S | |
| WSFQLUVWDKCYSW-UHFFFAOYNA-M | |
| [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O |
Safety and Handling
GHS H Statement Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only