missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MOPS-SDS running buffer (20X)

MOPS-SDS running buffer (20X) is a buffer frequently used for separating medium- to large-sized proteins on a gel.

Specifications

Chemical Name or Material MOPS-SDS running buffer
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ62847AP
View Documents Promotion Details
Thermo Scientific™
J62847AP
500mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ62847K2
View Documents Promotion Details
Thermo Scientific™
J62847K2
1L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • MOPS-SDS running buffer is a zwitterionic, morpholinic buffer that allows the separation of medium- to large-sized proteins

Application

  • MOPS is used as a running buffer in electrophoresis. The use of MOPS buffer allows proteins to run slower than when MES buffer is used
  • MOPS-SDS running buffer, 20X concentrated, is formulated to be used for running proteins on an SDS gel
  • This buffer contains SDS detergent to maintain the denaturation of proteins and MOPS to buffer the pH
  • It is commonly used for cell culture media and protein purification in chromatography
  • MOPS is frequently applied in buffered culture media for bacteria, yeast, and mammalian cells
  • It is also applied in the bicinchoninic acid (BCA) assay
  • It is used in separating RNA in agarose gels
Specifications

Specifications

MOPS-SDS running buffer
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only