missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosodium Risedronate Hemipentahydrate 98.0+%, TCI America™
$180.97 - $1028.05
Chemical Identifiers
| CAS | 329003-65-8 |
|---|---|
| Molecular Formula | C14H30N2Na2O19P4 |
| Molecular Weight (g/mol) | 700.264 |
| MDL Number | MFCD01706268 |
| InChI Key | HYFDYHPNTXOPPO-UHFFFAOYSA-L |
| Synonym | Monosodium 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonate, Risedronic Acid Monosodium Salt |
| PubChem CID | 11954322 |
| IUPAC Name | disodium;hydroxy-(1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphinate;pentahydrate |
| SMILES | C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].O.O.O.O.O.[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2289100MG
|
TCI America
M2289100MG |
100 mg |
Each for $180.97
|
|
|||||
|
M22891G
|
TCI America
M22891G |
1 g |
Each for $1,028.05
|
|
|||||
Chemical Identifiers
| 329003-65-8 | |
| 700.264 | |
| HYFDYHPNTXOPPO-UHFFFAOYSA-L | |
| 11954322 | |
| C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].O.O.O.O.O.[Na+].[Na+] |
| C14H30N2Na2O19P4 | |
| MFCD01706268 | |
| Monosodium 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonate, Risedronic Acid Monosodium Salt | |
| disodium;hydroxy-(1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphinate;pentahydrate |
Specifications
| 329003-65-8 | |
| C14H30N2Na2O19P4 | |
| 100 mg | |
| HYFDYHPNTXOPPO-UHFFFAOYSA-L | |
| disodium;hydroxy-(1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphinate;pentahydrate | |
| 11954322 | |
| ≥98.0% (HPLC,T) | |
| Monosodium Risedronate Hemipentahydrate |
| White | |
| MFCD01706268 | |
| Monosodium 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonate, Risedronic Acid Monosodium Salt | |
| C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].O.O.O.O.O.[Na+].[Na+] | |
| 700.264 | |
| 305.09 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : SZ8567500
TSCA : No
Recommended Storage : Refrigerator