missing translation for 'onlineSavingsMsg'
Learn More

Monosodium Risedronate Hemipentahydrate 98.0+%, TCI America™

Catalog No. M22891G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22891G 1 g
M2289100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22891G Supplier TCI America Supplier No. M22891G
Only null left

Chemical Identifiers

CAS 329003-65-8
Molecular Formula C14H30N2Na2O19P4
Molecular Weight (g/mol) 700.264
MDL Number MFCD01706268
InChI Key HYFDYHPNTXOPPO-UHFFFAOYSA-L
Synonym Monosodium 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonate, Risedronic Acid Monosodium Salt
PubChem CID 11954322
IUPAC Name disodium;hydroxy-(1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphinate;pentahydrate
SMILES C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].C1=CC(=CN=C1)CC(O)(P(=O)(O)O)P(=O)(O)[O-].O.O.O.O.O.[Na+].[Na+]

Specifications

Color White
Quantity 1 g
Formula Weight 305.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monosodium Risedronate Hemipentahydrate
RTECSNumber SZ8567500
TSCA No
Recommended Storage Refrigerator