missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosodium Risedronate Hemipentahydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 100 mg |
| Formula Weight | 305.09 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Monosodium Risedronate Hemipentahydrate |
Safety and Handling
| RTECSNumber | SZ8567500 |
| TSCA | No |
| Recommended Storage | Refrigerator |