missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monopotassium 7-Amino-1,3-naphthalenedisulfonate Hydrate 98.0+%, TCI America™
Supplier: TCI America A0337500G
Specifications
| Monopotassium 7-Amino-1,3-naphthalenedisulfonate Hydrate | |
| White-Yellow | |
| MFCD00064177 | |
| Amino G Acid Monopotassium Salt, 7-Amino-1,3-naphthalenedisulfonic Acid Monopotassium Salt, 2-Naphthylamine-6,8-disulfonic Acid Monopotassium Salt, Potassium Hydrogen 7-Amino-1,3-naphthalenedisulfonate | |
| C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N.[K] | |
| 342.401 | |
| 341.39 | |
| Crystalline Powder |
| 842-15-9 | |
| C10H9KNO6S2 | |
| 500 g | |
| QNHMTLCVDUMUFH-UHFFFAOYSA-N | |
| 7-aminonaphthalene-1,3-disulfonic acid;potassium | |
| 88146376 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 842-15-9 | |
| 342.401 | |
| QNHMTLCVDUMUFH-UHFFFAOYSA-N | |
| 88146376 | |
| C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N.[K] |
| C10H9KNO6S2 | |
| MFCD00064177 | |
| Amino G Acid Monopotassium Salt, 7-Amino-1,3-naphthalenedisulfonic Acid Monopotassium Salt, 2-Naphthylamine-6,8-disulfonic Acid Monopotassium Salt, Potassium Hydrogen 7-Amino-1,3-naphthalenedisulfonate | |
| 7-aminonaphthalene-1,3-disulfonic acid;potassium |
Safety and Handling
TSCA : Yes