missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomyristin 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 302.46 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Monomyristin |
Safety and Handling
| EINECSNumber | (2)-0669 |
| TSCA | No |