accessibility menu, dialog, popup

UserName

CAS RN 589-68-4

H3COOOHOH

CAS RN 589-68-4

IUPAC Name: 2,3-dihydroxypropyl tetradecanoate
Synonyms: 2,3-dihydroxypropyl tetradecanoate
Molecular Weight (g/mol): 302.46
Molecular Formula: C17H34O4
InChi Key: DCBSHORRWZKAKO-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCC(=O)OCC(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results