missing translation for 'onlineSavingsMsg'
Learn More

Monomyristin 97.0+%, TCI America™

Catalog No. M10731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10731G 1 g
M10735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10731G Supplier TCI America Supplier No. M10731G
Only null left

Chemical Identifiers

CAS 589-68-4
Molecular Formula C17H34O4
Molecular Weight (g/mol) 302.455
MDL Number MFCD00046760
InChI Key DCBSHORRWZKAKO-UHFFFAOYSA-N
Synonym Glycerol alpha-Monomyristate, 1-Monomyristoyl Glycerol
PubChem CID 79050
ChEBI CHEBI:75562
IUPAC Name 2,3-dihydroxypropyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(CO)O

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 1 g
Formula Weight 302.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monomyristin
EINECSNumber (2)-0669
TSCA No