missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl trans-1,4-Cyclohexanedicarboxylate 97.0+%, TCI America™

Catalog No. p-7133270
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25265G 5 g
M252625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25265G Supplier TCI America Supplier No. M25265G
Only null left

Chemical Identifiers

CAS 15177-67-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD01311247
InChI Key ZQJNPHCQABYENK-UHFFFAOYSA-N
Synonym trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester, Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate
PubChem CID 296850
IUPAC Name 4-methoxycarbonylcyclohexane-1-carboxylic acid
SMILES COC(=O)C1CCC(CC1)C(=O)O

Specifications

Melting Point 128°C
Color White
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl trans-1,4-Cyclohexanedicarboxylate