CAS RN 15177-67-0
IUPAC Name:
4-(methoxycarbonyl)cyclohexane-1-carboxylic acid
Synonyms:
4-(methoxycarbonyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
186.21
Molecular Formula:
C9H14O4
InChi Key:
ZQJNPHCQABYENK-UHFFFAOYSA-N
SMILES:
COC(=O)C1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
Monomethyl trans-1,4-Cyclohexanedicarboxylate 97.0+%, TCI America™
CAS: 15177-67-0 Molecular Formula: C9H14O4 Molecular Weight (g/mol): 186.207 MDL Number: MFCD01311247 InChI Key: ZQJNPHCQABYENK-UHFFFAOYSA-N Synonym: trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester, Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate PubChem CID: 296850 IUPAC Name: 4-methoxycarbonylcyclohexane-1-carboxylic acid SMILES: COC(=O)C1CCC(CC1)C(=O)O