missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl trans-1,4-Cyclohexanedicarboxylate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 128°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 186.21 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Monomethyl trans-1,4-Cyclohexanedicarboxylate |
Safety and Handling
| TSCA | No |