missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Terephthalate 98.0+%, TCI America™

Catalog No. p-7136161
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T024425G 25 g
T0244500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T024425G Supplier TCI America Supplier No. T024425G
Only null left

Chemical Identifiers

CAS 1679-64-7
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00002557
InChI Key REIDAMBAPLIATC-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl benzoic acid, mono-methyl terephthalate, monomethyl terephthalate, methyl terephthalate, methyl hydrogen terephthalate, 1,4-benzenedicarboxylic acid, monomethyl ester, terephthalic acid monomethyl ester, 1,4-benzenedicarboxylic acid monomethyl ester, terephthalic acid, monomethyl ester, hydrogen methyl terephthalate
PubChem CID 15513
IUPAC Name 4-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 222°C
Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl Terephthalate
EINECSNumber (3)-1326
TSCA Yes