missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl Maleate 90.0+%, TCI America™
Supplier: TCI America M0624100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| C5H6O4 | |
| MFCD00045905,MFCD00063174 | |
| Maleic Acid Monomethyl Ester, Methyl Hydrogen Maleate | |
| (2Z)-4-methoxy-4-oxobut-2-enoic acid |
Specifications
| Monomethyl Maleate | |
| Yellow | |
| 100 mL | |
| Maleic Acid Monomethyl Ester, Methyl Hydrogen Maleate | |
| COC(=O)\C=C/C(O)=O | |
| 130.10 | |
| 130.10 | |
| Liquid |
| 3052-50-4 | |
| C5H6O4 | |
| MFCD00045905,MFCD00063174 | |
| NKHAVTQWNUWKEO-IHWYPQMZSA-N | |
| (2Z)-4-methoxy-4-oxobut-2-enoic acid | |
| 5354456 | |
| ≥90.0% (GC) |
Safety and Handling
EINECSNumber : (2)-1105&(2)-1142
TSCA : Yes
Recommended Storage : Refrigerator