missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Maleate 90.0+%, TCI America™

$165.18 - $738.56

Chemical Identifiers

CAS 3052-50-4
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.10
MDL Number MFCD00045905,MFCD00063174
InChI Key NKHAVTQWNUWKEO-IHWYPQMZSA-N
Synonym Maleic Acid Monomethyl Ester, Methyl Hydrogen Maleate
PubChem CID 5354456
IUPAC Name (2Z)-4-methoxy-4-oxobut-2-enoic acid
SMILES COC(=O)\C=C/C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M062425ML
View Documents Promotion Details
N/A 25 mL
Each for $165.18
Only null left
 
M0624100ML
View Documents Promotion Details
N/A 100 mL
Each for $345.54
Only null left
 
M0624500ML
View Documents Promotion Details
N/A 500 mL
Each for $738.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3052-50-4
130.10
NKHAVTQWNUWKEO-IHWYPQMZSA-N
5354456
COC(=O)\C=C/C(O)=O
C5H6O4
MFCD00045905,MFCD00063174
Maleic Acid Monomethyl Ester, Methyl Hydrogen Maleate
(2Z)-4-methoxy-4-oxobut-2-enoic acid

Specifications

3052-50-4
C5H6O4
Maleic Acid Monomethyl Ester, Methyl Hydrogen Maleate
COC(=O)\C=C/C(O)=O
130.10
130.10
Liquid
Yellow
MFCD00045905,MFCD00063174
NKHAVTQWNUWKEO-IHWYPQMZSA-N
(2Z)-4-methoxy-4-oxobut-2-enoic acid
5354456
≥90.0% (GC)
Monomethyl Maleate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1105&(2)-1142

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents