missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl Adipate 97.0+%, TCI America™
$109.98 - $337.40
Chemical Identifiers
| CAS | 627-91-8 |
|---|---|
| Molecular Formula | C7H12O4 |
| Molecular Weight (g/mol) | 160.17 |
| MDL Number | MFCD00004418 |
| InChI Key | UOBSVARXACCLLH-UHFFFAOYSA-N |
| Synonym | monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester |
| PubChem CID | 12328 |
| ChEBI | CHEBI:70855 |
| IUPAC Name | 6-methoxy-6-oxohexanoic acid |
| SMILES | COC(=O)CCCCC(O)=O |
Chemical Identifiers
| 627-91-8 | |
| 160.17 | |
| UOBSVARXACCLLH-UHFFFAOYSA-N | |
| 12328 | |
| 6-methoxy-6-oxohexanoic acid |
| C7H12O4 | |
| MFCD00004418 | |
| monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester | |
| CHEBI:70855 | |
| COC(=O)CCCCC(O)=O |
Specifications
| 627-91-8 | |
| Yellow | |
| C7H12O4 | |
| 25 g | |
| UOBSVARXACCLLH-UHFFFAOYSA-N | |
| 6-methoxy-6-oxohexanoic acid | |
| 12328 | |
| 160.17 | |
| Liquid |
| 9°C | |
| 162°C | |
| MFCD00004418 | |
| monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester | |
| COC(=O)CCCCC(O)=O | |
| 160.17 | |
| CHEBI:70855 | |
| ≥97.0% (GC,T) | |
| Monomethyl Adipate |
Safety and Handling
EINECSNumber : (2)-0861
TSCA : Yes