accessibility menu, dialog, popup

UserName

CAS RN 627-91-8

IUPAC Name: 6-methoxy-6-oxohexanoic acid
Synonyms: hexanedioic acid, monomethyl ester
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: UOBSVARXACCLLH-UHFFFAOYSA-N
SMILES: COC(=O)CCCCC(O)=O
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)