missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl adipate, 97%

CAS: 627-91-8 | C7H12O4 | 160.17 g/mol

$197.38 - $342.79

Chemical Identifiers

CAS 627-91-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004418
InChI Key UOBSVARXACCLLH-UHFFFAOYSA-N
Synonym monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
PubChem CID 12328
ChEBI CHEBI:70855
IUPAC Name 6-methoxy-6-oxohexanoic acid
SMILES COC(=O)CCCCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102820250
SDS View Documents
Thermo Scientific Chemicals
102820250
25 g Glass bottle
Each for $197.38
Only null left
 
AC102821000
SDS View Documents
Thermo Scientific Chemicals
102821000
100 g Glass bottle
Each for $342.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

627-91-8
160.17
UOBSVARXACCLLH-UHFFFAOYSA-N
12328
6-methoxy-6-oxohexanoic acid
C7H12O4
MFCD00004418
monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
CHEBI:70855
COC(=O)CCCCC(O)=O

Specifications

627-91-8
100.0
Colorless
162.0°C (10.0 mmHg)
Authentic
Glass bottle
1.4390 to 1.4420
MFCD00004418
02, III, 1711
monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
COC(=O)CCCCC(O)=O
160.17
CHEBI:70855
97%
Monomethyl adipate, 98%
97.5
8.0°C to 9.0°C
1.0800g/mL
>109°C
97%
C7H12O4
HO2C(CH2)4CO2CH3
25 g
1.08
UOBSVARXACCLLH-UHFFFAOYSA-N
6-methoxy-6-oxohexanoic acid
12328
160.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 211-019-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only