missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl adipate, 97%

Catalog No. AC102820250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102820250 25 g Glass bottle
AC102821000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102820250 Supplier Thermo Scientific Chemicals Supplier No. 102820250

CAS: 627-91-8 | C7H12O4 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-91-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004418
InChI Key UOBSVARXACCLLH-UHFFFAOYSA-N
Synonym monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
PubChem CID 12328
ChEBI CHEBI:70855
IUPAC Name 6-methoxy-6-oxohexanoic acid
SMILES COC(=O)CCCCC(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.0°C to 9.0°C
Color Colorless
Density 1.0800g/mL
Boiling Point 162.0°C (10.0 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4390 to 1.4420
Linear Formula HO2C(CH2)4CO2CH3
Quantity 25 g
Beilstein 02, III, 1711
Specific Gravity 1.08
Formula Weight 160.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Monomethyl adipate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 211-019-0
TSCA TSCA

RUO – Research Use Only