missing translation for 'onlineSavingsMsg'
Learn More

Covidien Monoject™ Tube Caps

Catalog No. 0254427
Change view
Click to view available options
Color:
Red
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0254427 White
0254423 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0254427 Supplier Covidien Supplier No. Z1270019500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy-on, easy-off caps for glass blood collection tubes

  • Exclusive thumb tab makes cap removal simple, quick and fatigue-free
  • Supplied in convenient, resealable and polybag-lined dispenser box
  • Ideal for benchtop use
  • Available in six colors to match the original blood tube stopper
  • Size: 13mm

Specifications

Color White
For Use With (Equipment) 13 mm glass blood collection tubes